Order Entry
Export
ContactUsLinkComponent
Anti-c-Myc Rabbit Recombinant Antibody [clone: MYC2895R]
Anti-c-Myc Rabbit Recombinant Antibody [clone: MYC2895R]
Catalog # ANTIA249457-100
Supplier:  Antibodies.com
New Product
undefined
Anti-c-Myc Rabbit Recombinant Antibody [clone: MYC2895R]
Catalog # ANTIA249457-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AU016757
  • Antigen symbol:
    c-Myc
  • Clonality:
    Recombinant
  • Clone:
    MYC2895R
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P01106
  • Antigen synonyms:
    MRTL|Class E basic helix-loop-helix protein 39|bHLHe39|c Myc|Avian myelocytomatosis viral oncogene homolog|c-Myc|Myc Epitope Tag|Myc protein|Myc proto oncogene protein
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    62 - 64 kDa
  • Sequence:
    AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to amino acids 408-439 of human c-Myc, coupled to KLH
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [MYC2895R] antibody to c-Myc for ELISA, flow cytometry and IF with samples derived from human.

  • Validated applications: ELISA, Flow cytometry, IF
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: c-Myc
Clonality: Recombinant
Clone: MYC2895R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human