Order Entry
Export
ContactUsLinkComponent
 
Anti-c-Myc Mouse Monoclonal Antibody [clone: MYC275 + MYC909]
  ANTIA249454-100
New Product
undefined
Anti-c-Myc Mouse Monoclonal Antibody [clone: MYC275 + MYC909]
  ANTIA249454-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    AU016757
  • Antigen symbol:
    c-Myc
  • Clonality:
    Monoclonal
  • Clone:
    MYC275 + MYC909
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P01106
  • Antigen synonyms:
    MRTL|Class E basic helix-loop-helix protein 39|bHLHe39|c Myc|Avian myelocytomatosis viral oncogene homolog|c-Myc|Myc Epitope Tag|Myc protein|Myc proto oncogene protein
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    62 - 64 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human c-Myc protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [MYC275 + MYC909] antibody to c-Myc for flow cytometry, IF and IHC-P with samples derived from human.

  • Validated applications: Flow cytometry, IF, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: c-Myc
Clonality: Monoclonal
Clone: MYC275 + MYC909
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human