Anti-c-Myc Mouse Monoclonal Antibody [clone: MYC909]
Catalog # ANTIA249453-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:AU016757
- Antigen symbol:c-Myc
- Clonality:Monoclonal
- Clone:MYC909
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P01106
- Antigen synonyms:MRTL|Class E basic helix-loop-helix protein 39|bHLHe39|c Myc|Avian myelocytomatosis viral oncogene homolog|c-Myc|Myc Epitope Tag|Myc protein|Myc proto oncogene protein
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:62 - 64 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human c-Myc protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [MYC909] antibody to c-Myc for flow cytometry, IF and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: c-Myc
Clonality: Monoclonal
Clone: MYC909
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human