Order Entry
Export
ContactUsLinkComponent
Anti-LSP1 Mouse Monoclonal Antibody [clone: LSP1/3042]
Anti-LSP1 Mouse Monoclonal Antibody [clone: LSP1/3042]
  ANTIA249238-100
New Product
undefined
Anti-LSP1 Mouse Monoclonal Antibody [clone: LSP1/3042]
  ANTIA249238-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    47 kDa actin binding protein
  • Antigen symbol:
    LSP1
  • Clonality:
    Monoclonal
  • Clone:
    LSP1/3042
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P33241
  • Antigen synonyms:
    Leufactin|LSP 1|52 kDa phosphoprotein|Leukocyte F actin binding protein|47 kDa actin-binding protein|LSP1|F actin binding and cytoskeleton associated protein|Leukocyte specific protein 1|Leufactin (leukocyte F-actin binding protein)
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    52 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 174-278, of human LSP1 protein. The exact sequence is proprietary.
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [LSP1/3042] antibody to LSP1 for ELISA with samples derived from human.

  • Validated applications: ELISA

Type: Primary
Antigen: LSP1
Clonality: Monoclonal
Clone: LSP1/3042
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human