Anti-LSP1 Mouse Monoclonal Antibody [clone: LSP1/3025]
ANTIA249237-100
New Product
- Antibody type:Primary
- Antigen name:47 kDa actin binding protein
- Antigen symbol:LSP1
- Clonality:Monoclonal
- Clone:LSP1/3025
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P33241
- Antigen synonyms:Leufactin|LSP 1|52 kDa phosphoprotein|Leukocyte F actin binding protein|47 kDa actin-binding protein|LSP1|F actin binding and cytoskeleton associated protein|Leukocyte specific protein 1|Leufactin (leukocyte F-actin binding protein)
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:52 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 174-278, of human LSP1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [LSP1/3025] antibody to LSP1 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: LSP1
Clonality: Monoclonal
Clone: LSP1/3025
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human