Order Entry
Export
ContactUsLinkComponent
Anti-Cytokeratin 19 Mouse Monoclonal Antibody [clone: KRT19/799]
Anti-Cytokeratin 19 Mouse Monoclonal Antibody [clone: KRT19/799]
Catalog # ANTIA249213-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Cytokeratin 19 Mouse Monoclonal Antibody [clone: KRT19/799]
Catalog # ANTIA249213-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    40 kDa keratin intermediate filament
  • Antigen symbol:
    Cytokeratin 19
  • Clonality:
    Monoclonal
  • Clone:
    KRT19/799
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a lambda
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P08727
  • Antigen synonyms:
    Cytokeratin-19|CK19|CK-19|K1CS|Keratin type I 40kD|Cytokeratin 19|CK 19|Keratin type I 40 kD|K1C19_HUMAN
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    40 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human KRT19 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [KRT19/799] antibody to Cytokeratin 19 for flow cytometry, IF, WB and IHC-P with samples derived from human.

  • Validated applications: Flow cytometry, IF, WB, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 19
Clonality: Monoclonal
Clone: KRT19/799
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a lambda
Reactivity: Human