Anti-Cytokeratin 18 Rabbit Recombinant Antibody [clone: KRT18/2819R]
Catalog # ANTIA249206-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cell proliferation inducing gene 46 protein
- Antigen symbol:Cytokeratin 18
- Clonality:Recombinant
- Clone:KRT18/2819R
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse,Cat,Marmoset,Hamster,Pig,Horse,Sheep,Goat,Ferret,Human,Bovine,Dog,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P05783
- Antigen synonyms:CK18|Cell proliferation inducing protein 46|Cytokeratin 18|CYK18|Cytokeratin endo B|Cell proliferation-inducing gene 46 protein|CK 18|CK-18|CYK 18
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:45 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human KRT18 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [KRT18/2819R] antibody to Cytokeratin 18 for flow cytometry, WB, IF and IHC-P with samples derived from human, mouse, rat, hamster, ferret, bovine, porcine, feline, canine, sheep, goat, horse and Marmoset monkey.
- Validated applications: Flow cytometry, WB, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ml, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 18
Clonality: Recombinant
Clone: KRT18/2819R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster, Ferret, Bovine, Porcine, Cat, Dog, Sheep, Goat, Horse, Marmoset Monkey