Order Entry
Export
ContactUsLinkComponent
 
Anti-Cytokeratin 18 Mouse Monoclonal Antibody [clone: DC10]
  ANTIA249194-100
New Product
undefined
Anti-Cytokeratin 18 Mouse Monoclonal Antibody [clone: DC10]
  ANTIA249194-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Cell proliferation inducing gene 46 protein
  • Antigen symbol:
    Cytokeratin 18
  • Clonality:
    Monoclonal
  • Clone:
    DC10
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05783
  • Antigen synonyms:
    CK18|Cell proliferation inducing protein 46|Cytokeratin 18|CYK18|Cytokeratin endo B|Cell proliferation-inducing gene 46 protein|CK 18|CK-18|CYK 18
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    45 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    PMC-42 human breast carcinoma cells
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [DC10] antibody to Cytokeratin 18 for flow cytometry, IF, WB and IHC-P with samples derived from human.

  • Validated applications: Flow cytometry, IF, WB, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 18
Clonality: Monoclonal
Clone: DC10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human