Order Entry
Export
ContactUsLinkComponent
Anti-Cytokeratin 17 Mouse Monoclonal Antibody [clone: KRT17/778]
Anti-Cytokeratin 17 Mouse Monoclonal Antibody [clone: KRT17/778]
Catalog # ANTIA249191-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Cytokeratin 17 Mouse Monoclonal Antibody [clone: KRT17/778]
Catalog # ANTIA249191-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    keratin 17 epitope S2
  • Antigen symbol:
    Cytokeratin 17
  • Clonality:
    Monoclonal
  • Clone:
    KRT17/778
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Pig,
    Goat,
    Human,
    Bovine,
    Rat
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q04695
  • Antigen synonyms:
    CK-17|Cytokeratin 17|Keratin|39.1|K1C17_HUMAN|keratin 17 epitope S1|Cytokeratin-17|K17|CK 17
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    46 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human KRT17 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [KRT17/778] antibody to Cytokeratin 17 for IHC-P with samples derived from human, rat, bovine, goat and porcine.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 17
Clonality: Monoclonal
Clone: KRT17/778
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Rat, Bovine, Goat, Porcine