Anti-Cytokeratin 16 Mouse Monoclonal Antibody [clone: KRT16/1714]
Catalog # ANTIA249185-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CK 16
- Antigen symbol:Cytokeratin 16
- Clonality:Monoclonal
- Clone:KRT16/1714
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P08779
- Antigen synonyms:K1CP|Focal non epidermolytic palmoplantar keratoderma|FNEPPK|Cytokeratin 16|CK16|CK-16|Cytokeratin16|K1C16_HUMAN|Cytokeratin-16
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:48 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment from the C terminal of human Cytokeratin 16
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [KRT16/1714] antibody to Cytokeratin 16 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 16
Clonality: Monoclonal
Clone: KRT16/1714
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human