Anti-Cytokeratin 8 Mouse Monoclonal Antibody [clone: C-43]
Catalog # ANTIA249154-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CARD2
- Antigen symbol:Cytokeratin 8
- Clonality:Monoclonal
- Clone:C-43
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Pig,Rabbit,Sheep,Human,Bovine
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P05787
- Antigen synonyms:Cytokeratin endo A|CYK8|CK-8|CK 8|DreK8|Cytokeratin-8|CYKER|CK8|Cytokeratin 8
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:52,5 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Cytoskeletal preparation from HeLa cells
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [C-43] antibody to Cytokeratin 8 for IF, WB and IHC-P with samples derived from human, sheep, rabbit, bovine and porcine.
- Validated applications: IF, WB, IHC-P
- Recommended dilutions: IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 8
Clonality: Monoclonal
Clone: C-43
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Sheep, Rabbit, Bovine, Porcine