Order Entry
Export
ContactUsLinkComponent
Anti-Cytokeratin 7 Mouse Recombinant Antibody [clone: rOV-TL12/30]
Anti-Cytokeratin 7 Mouse Recombinant Antibody [clone: rOV-TL12/30]
Catalog # ANTIA249133-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Cytokeratin 7 Mouse Recombinant Antibody [clone: rOV-TL12/30]
Catalog # ANTIA249133-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CK 7
  • Antigen symbol:
    Cytokeratin 7
  • Clonality:
    Recombinant
  • Clone:
    rOV-TL12/30
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P08729
  • Antigen synonyms:
    CK7|K2C7|D15Wsu77e|Cytokeratin 7|Cytokeratin-7|Keratin 7, type II|CK-7|K2C7_HUMAN|Keratin type II cytoskeletal 7
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    55 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human KRT7 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant mouse monoclonal [rOV-TL12/30] antibody to Cytokeratin 7 for WB, flow cytometry and IHC-P with samples derived from human.

  • Validated applications: WB, Flow cytometry, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, Flow cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 7
Clonality: Recombinant
Clone: rOV-TL12/30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human