Anti-Integrin beta 3 Mouse Recombinant Antibody [clone: rITGB3/2145]
Catalog # ANTIA249084-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:BDPLT16
- Antigen symbol:Integrin beta 3
- Clonality:Recombinant
- Clone:rITGB3/2145
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P05106
- Antigen synonyms:HPA 1|CD61 antigen|GP3A|BDPLT2|CD 61|HPA 4|GPIIIa|GT|CD61
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:105 kDa/90 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 385-490, of human ITGB3 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rITGB3/2145] antibody to Integrin beta 3 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Integrin beta 3
Clonality: Recombinant
Clone: rITGB3/2145
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human