Anti-Lambda Light Chain Mouse Monoclonal Antibody [clone: SPM559]
ANTIA248994-100
New Product
- Antibody type:Primary
- Antigen name:Bence Jones protein
- Antigen symbol:Lambda Light Chain
- Clonality:Monoclonal
- Clone:SPM559
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P01701, P01842
- Antigen synonyms:Ig lambda chain C regions|BJP|IGLC 3|ig lambda-6 chain C region|IGLC 1|IGLC|IGLC 2|IGLC1|Constant region of lambda light chains
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:~22,5 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Purified human IgG protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [SPM559] antibody to Lambda Light Chain for flow cytometry, IF, WB and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Lambda Light Chain
Clonality: Monoclonal
Clone: SPM559
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human