Anti-Kappa Light Chain Mouse Monoclonal Antibody [clone: Kap-56]
Catalog # ANTIA248983-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:HCAK 1
- Antigen symbol:Kappa Light Chain
- Clonality:Monoclonal
- Clone:Kap-56
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P01601, P01834
- Antigen synonyms:immunoglobulin kappa constant|immunoglobulin kappa light chain constant region|immunoglobulin kappa light chain|IGKC|immunoglobulin InV|IGKCD|Ig kappa chain C region|HCAK1|immunoglobulin kappa constant region
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:~22,5 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human lymphocytes stimulated
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [Kap-56] antibody to Kappa Light Chain for flow cytometry, WB and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, WB, IHC-P
- Recommended dilutions: Flow cytometry: 0,5 to 2 µg/million cells, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Kappa Light Chain
Clonality: Monoclonal
Clone: Kap-56
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human