Order Entry
Export
ContactUsLinkComponent
 
Anti-Kappa Light Chain Mouse Recombinant Antibody [clone: rKLC264]
  ANTIA248973-100
New Product
undefined
Anti-Kappa Light Chain Mouse Recombinant Antibody [clone: rKLC264]
  ANTIA248973-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    HCAK 1
  • Antigen symbol:
    Kappa Light Chain
  • Clonality:
    Recombinant
  • Clone:
    rKLC264
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P01601, P01834
  • Antigen synonyms:
    immunoglobulin kappa constant|immunoglobulin kappa light chain constant region|immunoglobulin kappa light chain|IGKC|immunoglobulin InV|IGKCD|Ig kappa chain C region|HCAK1|immunoglobulin kappa constant region
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    ~22,5 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human Ig kappa chain
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant mouse monoclonal [rKLC264] antibody to Kappa Light Chain for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Kappa Light Chain
Clonality: Recombinant
Clone: rKLC264
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human