Order Entry
Export
ContactUsLinkComponent
Anti-ICAM1 Mouse Monoclonal Antibody [clone: P2A4]
Anti-ICAM1 Mouse Monoclonal Antibody [clone: P2A4]
Catalog # ANTIA248905-100
Supplier:  Antibodies.com
New Product
undefined
Anti-ICAM1 Mouse Monoclonal Antibody [clone: P2A4]
Catalog # ANTIA248905-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Antigen identified by monoclonal
  • Antigen symbol:
    ICAM1
  • Clonality:
    Monoclonal
  • Clone:
    P2A4
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05362
  • Antigen synonyms:
    Human rhinovirus receptor|CD54 antigen|Cell surface glycoprotein P3.58|BB2|CD54|CD_antigen=CD54|BB 2|CD 54|ICAM 1
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    85 - 115 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human ICAM1 protein. Epitope is the extracellular domain.
  • Tested applications:
    Functional studies, IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [P2A4] antibody to ICAM1 for functional studies, flow cytometry, IF and IHC-P with samples derived from human.

  • Validated applications: Functional studies, Flow cytometry, IF, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: P2A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human