Anti-ICAM1 Mouse Monoclonal Antibody [clone: F4-31C2]
Catalog # ANTIA248903-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antigen identified by monoclonal
- Antigen symbol:ICAM1
- Clonality:Monoclonal
- Clone:F4-31C2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P05362
- Antigen synonyms:Human rhinovirus receptor|CD54 antigen|Cell surface glycoprotein P3.58|BB2|CD54|CD_antigen=CD54|BB 2|CD 54|ICAM 1
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:85 - 115 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human umbilical vein endothelial cells (HUVEC)
- Tested applications:Functional studies
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [F4-31C2] antibody to ICAM1 for functional studies, flow cytometry and IF with samples derived from human.
- Validated applications: Functional studies, Flow cytometry, IF
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: F4-31C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human