Anti-HSP27 Mouse Monoclonal Antibody [clone: SPM252]
ANTIA248873-100
New Product
- Antibody type:Primary
- Antigen name:28 kDa heat shock protein
- Antigen symbol:HSP27
- Clonality:Monoclonal
- Clone:SPM252
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Sheep,Chicken,Chimpanzee,Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P04792
- Antigen synonyms:epididymis secretory protein Li 102|Heat shock 27 kDa protein|Estrogen regulated 24 kDa protein|Heat shock 27kD protein 1|Heat shock 27kDa protein|Estrogen-regulated 24 kDa protein|DKFZp586P1322|CMT2F|Heat shock 25kDa protein 1
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:27 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Partially purified HSP27 (originally called 24K) protein from breast cancer MCF-7 cells
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [SPM252] antibody to HSP27 for flow cytometry, IF, WB and IHC-P with samples derived from human, chimpanzee, Sheep, rat, mouse and chicken.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: HSP27
Clonality: Monoclonal
Clone: SPM252
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Chimpanzee, Sheep, Rat, Mouse, Chicken