Order Entry
Export
ContactUsLinkComponent
Anti-HLA DRA Mouse Monoclonal Antibody [clone: 19-26.1]
Anti-HLA DRA Mouse Monoclonal Antibody [clone: 19-26.1]
  ANTIA248849-100
New Product
undefined
Anti-HLA DRA Mouse Monoclonal Antibody [clone: 19-26.1]
  ANTIA248849-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    DR alpha chain
  • Antigen symbol:
    HLA DRA
  • Clonality:
    Monoclonal
  • Clone:
    19-26.1
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P01903
  • Antigen synonyms:
    DR alpha chain precursor|HLA DR1B|DRB1|HLA DR3B|HLA class II histocompatibility antigen DR alpha chain|Histocompatibility antigen HLA DR alpha|DRA_HUMAN|HLA class II histocompatibility antigen|DRB4
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    36 kDa (alpha chain)/27 kDa (beta chain)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Raji cells
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [19-26.1] antibody to HLA DRA for flow cytometry, IF and IHC-P with samples derived from human.

  • Validated applications: Flow cytometry, IF, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: HLA DRA
Clonality: Monoclonal
Clone: 19-26.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human