Anti-HLA DP + DQ + DR Mouse Monoclonal Antibody [clone: CR3/43]
Catalog # ANTIA248841-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen symbol:HLA DP + DQ + DR
- Clonality:Monoclonal
- Clone:CR3/43
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P04440, P01908, P01909, P01920, P01903
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:36 kDa (alpha chain)/27 kDa (beta chain)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Cells from human tonsil
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [CR3/43] antibody to HLA DP + DQ + DR for ELISA, flow cytometry, IF, WB and IHC-P with samples derived from human.
- Validated applications: ELISA, Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: ELISA: 2 to 4 µg/ml, Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: HLA DP + DQ + DR
Clonality: Monoclonal
Clone: CR3/43
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human