Anti-CD57 Mouse Monoclonal Antibody [clone: SPM129]
Catalog # ANTIA248724-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:3-glucuronyltransferase
- Antigen symbol:CD57
- Clonality:Monoclonal
- Clone:SPM129
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q9P2W7
- Antigen synonyms:Beta 1 3 glucuronyltransferase 1|3-glucuronyltransferase 1|CD57|Beta-1|CD 57|B3gat1|CD57 antigen|Beta-1,3-glucuronyltransferase 1|B3GA1_HUMAN
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:~110 kDa (Glycoprotein)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Membrane antigen from HSB-2 cells
- Tested applications:IHC-P
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [SPM129] antibody to CD57 for IF and IHC-P with samples derived from human.
- Validated applications: IF, IHC-P
- Recommended dilutions: IF: 1 to 2 µg/ml, IHC-P: 2 to 4 µg/ml
Type: Primary
Antigen: CD57
Clonality: Monoclonal
Clone: SPM129
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human