Order Entry
Export
ContactUsLinkComponent
Anti-Connexin 32 Mouse Monoclonal Antibody [clone: GJB1/1753]
Anti-Connexin 32 Mouse Monoclonal Antibody [clone: GJB1/1753]
Catalog # ANTIA248719-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Connexin 32 Mouse Monoclonal Antibody [clone: GJB1/1753]
Catalog # ANTIA248719-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Connexin 32
  • Clonality:
    Monoclonal
  • Clone:
    GJB1/1753
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P08034
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    27 - 32 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human GJB1 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [GJB1/1753] antibody to Connexin 32 for IHC-P with samples derived from human, mouse and rat.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Connexin 32
Clonality: Monoclonal
Clone: GJB1/1753
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat