Anti-Albumin Mouse Monoclonal Antibody [clone: ALB/2142]
Catalog # ANTIA248499-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Growth inhibiting protein 20
- Antigen symbol:Albumin
- Clonality:Monoclonal
- Clone:ALB/2142
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P02768
- Antigen synonyms:GIG20|DKFZp779N1935|ALBU_HUMAN|alb|GIG42|Albumin (32 AA)|cell growth inhibiting protein 42|Albumin (AA 34)|Albumin
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:66 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human ALB protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [ALB/2142] antibody to Albumin for ELISA and IHC-P with samples derived from human.
- Validated applications: ELISA, IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Albumin
Clonality: Monoclonal
Clone: ALB/2142
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human