Order Entry
Export
ContactUsLinkComponent
Anti-Desmoglein 1 Mouse Monoclonal Antibody [clone: 32-2B]
Anti-Desmoglein 1 Mouse Monoclonal Antibody [clone: 32-2B]
Catalog # ANTIA248369-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Desmoglein 1 Mouse Monoclonal Antibody [clone: 32-2B]
Catalog # ANTIA248369-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cadherin family member 4
  • Antigen symbol:
    Desmoglein 1
  • Clonality:
    Monoclonal
  • Clone:
    32-2B
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q02413
  • Antigen synonyms:
    DGI|DG1|DSG1_HUMAN|Desmoglein 1|DSG1|Desmosomal glycoprotein 1|CDHF4|Desmoglein-1|DSG
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    150 kDa (precursor)/160 kDa (mature)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Bovine desmoglein (desmosomal glyco protein I) from nasal epidermis
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [32-2B] antibody to Desmoglein 1 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Desmoglein 1
Clonality: Monoclonal
Clone: 32-2B
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human