Order Entry
Export
ContactUsLinkComponent
Anti-CD55 Mouse Monoclonal Antibody [clone: 143-30]
Anti-CD55 Mouse Monoclonal Antibody [clone: 143-30]
Catalog # ANTIA248317-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD55 Mouse Monoclonal Antibody [clone: 143-30]
Catalog # ANTIA248317-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD 55
  • Antigen symbol:
    CD55
  • Clonality:
    Monoclonal
  • Clone:
    143-30
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P08174
  • Antigen synonyms:
    Complement decay-accelerating factor|CD55 Cromer blood group system|Complement decay accelerating factor|CD55 molecule, decay accelerating factor for complement (Cromer blood group)|CD55 antigen|CD55 molecule|CD55 molecule (Cromer blood group)|CD55|Cd55a
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    70 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    PHA stimulated human PBL
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [143-30] antibody to CD55 for flow cytometry and IF with samples derived from human.

  • Validated applications: Flow cytometry, IF
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: 143-30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human