Order Entry
Export
ContactUsLinkComponent
Anti-GM-CSF Rat Monoclonal Antibody [clone: SPM469]
Anti-GM-CSF Rat Monoclonal Antibody [clone: SPM469]
Catalog # ANTIA248274-100
Supplier:  Antibodies.com
New Product
Anti-GM-CSF Rat Monoclonal Antibody [clone: SPM469]
Catalog # ANTIA248274-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Colony stimulating factor
  • Antigen symbol:
    GM-CSF
  • Clonality:
    Monoclonal
  • Clone:
    SPM469
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Cynomolgus monkey,
    Rhesus monkey,
    Human
  • Neutralisation:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P04141
  • Antigen synonyms:
    CSF2_HUMAN|GMCSF|Colony-stimulating factor|Colony Stimulating Factor 2|GM-CSF|Colony stimulating factor 2 (granulocyte-macrophage)|CSF2|CSF 2|CSF
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    22 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human GM-CSF protein
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rat monoclonal [SPM469] antibody to GM-CSF for neutralization studies, flow cytometry and IF with samples derived from human, cynomolgus monkey and rhesus monkey.

  • Validated applications: Neutralization Studies, Flow cytometry, IF
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: GM-CSF
Clonality: Monoclonal
Clone: SPM469
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human, Cynomolgus Monkey, Rhesus Monkey