Order Entry
Export
ContactUsLinkComponent
Anti-TOP1MT Mouse Monoclonal Antibody [clone: TOP1MT/613]
Anti-TOP1MT Mouse Monoclonal Antibody [clone: TOP1MT/613]
Catalog # ANTIA248223-100
Supplier:  Antibodies.com
New Product
undefined
Anti-TOP1MT Mouse Monoclonal Antibody [clone: TOP1MT/613]
Catalog # ANTIA248223-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    2900052H09Rik
  • Antigen symbol:
    TOP1MT
  • Clonality:
    Monoclonal
  • Clone:
    TOP1MT/613
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q969P6
  • Antigen synonyms:
    DNA topoisomerase I, mitochondrial|TOP1M_HUMAN|Topoisomerase I mitochondrial|TOP1MT|DNA topoisomerase I
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    70 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human TOP1MT protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [TOP1MT/613] antibody to TOP1MT for WB and IHC-P with samples derived from human.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: TOP1MT
Clonality: Monoclonal
Clone: TOP1MT/613
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human