Anti-NACC1 Mouse Monoclonal Antibody [clone: PCRP-NACC1-1A8]
Catalog # ANTIA248213-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:BEND8
- Antigen symbol:NACC1
- Clonality:Monoclonal
- Clone:PCRP-NACC1-1A8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q96RE7
- Antigen synonyms:Nacc1|BTB/POZ domain-containing protein 14B|FLJ37383|BTB Domain Containing 14B|btbd14b|NAC-1|NACC1_HUMAN|Nac1|Nucleus accumbens-associated protein 1
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:57 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human NACC1 protein
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [PCRP-NACC1-1A8] antibody to NACC1 for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells
Type: Primary
Antigen: NACC1
Clonality: Monoclonal
Clone: PCRP-NACC1-1A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human