Anti-HCG beta Rabbit Recombinant Antibody [clone: HCGb/1996R]
ANTIA248175-100
New Product
- Antibody type:Primary
- Antigen name:CG beta
- Antigen symbol:HCG beta
- Clonality:Recombinant
- Clone:HCGb/1996R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P01233
- Antigen synonyms:CGB3|Chorionic gonadotrophin chain beta|CGHB_HUMAN|CG-beta|Choriogonadotropin subunit beta|CGB8|CGB5|CGB|CGB7
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:22 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length hCG beta protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [HCGb/1996R] antibody to HCG beta for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: HCG beta
Clonality: Recombinant
Clone: HCGb/1996R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human