Order Entry
Export
ContactUsLinkComponent
 
Anti-HCG beta Mouse Monoclonal Antibody [clone: HCGb/54+ HCGb/459]
  ANTIA248172-100
New Product
undefined
Anti-HCG beta Mouse Monoclonal Antibody [clone: HCGb/54+ HCGb/459]
  ANTIA248172-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    CG beta
  • Antigen symbol:
    HCG beta
  • Clonality:
    Monoclonal
  • Clone:
    HCGb/54+ HCGb/459
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P01233
  • Antigen synonyms:
    CGB3|Chorionic gonadotrophin chain beta|CGHB_HUMAN|CG-beta|Choriogonadotropin subunit beta|CGB8|CGB5|CGB|CGB7
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    22 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant hCG beta protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [HCGb/54+ HCGb/459] antibody to HCG beta for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: HCG beta
Clonality: Monoclonal
Clone: HCGb/54+ HCGb/459
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human