Order Entry
Export
ContactUsLinkComponent
Anti-HCG alpha Mouse Monoclonal Antibody [clone: HCGa/53]
Anti-HCG alpha Mouse Monoclonal Antibody [clone: HCGa/53]
Catalog # ANTIA248164-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-HCG alpha Mouse Monoclonal Antibody [clone: HCGa/53]
Catalog # ANTIA248164-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Anterior pituitary glycoprotein hormones Common subunit alpha
  • Antigen symbol:
    HCG alpha
  • Clonality:
    Monoclonal
  • Clone:
    HCGa/53
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P01215
  • Antigen synonyms:
    Follicle-stimulating hormone alpha|Chorionic Gonadotrophin Alpha Subunit|CG alpha|Chorionic Gonadotrophin Alpha|Choriogonadotropin alpha chain|CG-alpha|cga|Chorionic gonadotrophin subunit alpha|chorionic gonadotropin, alpha polypeptide
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    ~13 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant hCG alpha protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [HCGa/53] antibody to HCG alpha for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: HCG alpha
Clonality: Monoclonal
Clone: HCGa/53
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human