Anti-Cadherin 16 Mouse Monoclonal Antibody [clone: SPM594]
Catalog # ANTIA248066-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CAD16_HUMAN
- Antigen symbol:Cadherin 16
- Clonality:Monoclonal
- Clone:SPM594
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Rabbit,Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O75309
- Antigen synonyms:CDH16|Cadherin-16|CDH 16|Kidney specific cadherin|Cadherin 16 KSP cadherin|Cadherin 16|Kidney cadherin|Cadherin kidney|Kidney-specific cadherin
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:130 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human CDH16 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [SPM594] antibody to Cadherin 16 for WB and IHC-P with samples derived from human, mouse, rat, rabbit and canine.
- Validated applications: WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cadherin 16
Clonality: Monoclonal
Clone: SPM594
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Rabbit, Dog