Anti-CD172a Mouse Monoclonal Antibody [clone: 15-414] (PE (Phycoerythrin))
ANTIA242911-100
New Product
- Antibody type:Primary
- Antigen name:Bit
- Antigen symbol:CD172a
- Clonality:Monoclonal
- Clone:15-414
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P78324
- Antigen synonyms:CD172a|Brain immunoglobulin like molecule with tyrosine based activation motifs|CD172 antigen-like family member A|Inhibitory receptor SHPS-1|Brain Ig like molecule with tyrosine based activation motifs|Brain Ig-like molecule with tyrosine-based activation motifs|CD172a antigen|CD172 antigen like family member A|Macrophage fusion receptor
- Storage buffer:Supplied in phosphate buffered saline with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Kg-1a cell line
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Mouse monoclonal [15-414] antibody to CD172a (PE) for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD172a
Clonality: Monoclonal
Clone: 15-414
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human