Anti-B2M Mouse Monoclonal Antibody [clone: KT134]
Supplier: Antibodies.com
New Product
Certificates
About this item
Mouse monoclonal (KT134) antibody to B2M for ELISA, WB and sELISA with samples derived from human.
- Validated applications: ELISA, WB, sELISA
- Recommended dilutions: ELISA: 1/1000, WB: 1/500 to 1/10000, sELISA: 3 µg/ml
Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: KT134
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- ANTIA185-200
- ANTIA185-100
- Antigen symbol:
- B2M
- B2M
- Antigen synonyms:
- CDABP0092|Hdcma22p|Beta chain of mhc class 1 proteins|Beta chain of MHC class I molecules|Beta 2 microglobulin precursor|beta 2 Microglobulin|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin|B2MG_HUMAN
- CDABP0092|Hdcma22p|Beta chain of mhc class 1 proteins|Beta chain of MHC class I molecules|Beta 2 microglobulin precursor|beta 2 Microglobulin|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin|B2MG_HUMAN
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- KT134
- KT134
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P61769
- UniprotID# P61769
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Purified native human B2M
- Purified native human B2M
- Form:
- Liquid
- Liquid
- Purification:
- IgG is purified through a Protein A column
- IgG is purified through a Protein A column
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- Supplied in phosphate buffered saline with 0,09% sodium azide
- Supplied in phosphate buffered saline with 0,09% sodium azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Tested applications:
- Sandwich ELISA
- Sandwich ELISA