Order Entry
Export
ContactUsLinkComponent
Anti-ABCD4 Rabbit Polyclonal Antibody
Anti-ABCD4 Rabbit Polyclonal Antibody
Catalog # ANTIA305629-100
Supplier:  Antibodies.com
New Product
undefined
Anti-ABCD4 Rabbit Polyclonal Antibody
Catalog # ANTIA305629-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    69 kDa peroxisomal ABC transporter
  • Antigen symbol:
    ABCD4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O14678
  • Antigen synonyms:
    ABC41|ABC 41|EST352188|ABCD 4|ATP binding cassette sub family D member 4|ATP-binding cassette sub-family D member 4|ABCD4_HUMAN|ATP binding cassette sub family D (ALD) member 4|ABCD4
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% thiomersal
  • Molecular weight:
    72 kDa
  • Sequence:
    DQFTCNLLYVSWRKDLTEHLHRLYFRGRAYYTLNVLRDDIDNPDQRISQDVERFCRQLSSMASKLIISPFTLVYYTYQCFQSTGWLGPVSIFGYFILGTVVNKTLMGPIVMKLVHQEKLEGDFRFKHMQIRVNAEPAAFYRAGHVEHMRTDRRLQRLLQTQRELMSKELWLYIGINTFDYL
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 100 - 280 of human ABCD4 (NP_005041.1).
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to ABCD4 for WB with samples derived from human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: ABCD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human