Anti-TrkB Rabbit Monoclonal Antibody [clone: ARC53486]
Catalog # ANTIA305512-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:AI848316
- Antigen symbol:TrkB
- Clonality:Monoclonal
- Clone:ARC53486
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q16620
- Antigen synonyms:GP145-TrkB/GP95-TrkB|BDNF/NT-3 growth factors receptor|EC 2.7.10.1|BDNF/NT 3 growth factors receptor|GP145-TrkB|GP145 TrkB|C030027L06Rik|BDNF tropomyosine receptor kinase B|Brain derived neurotrophic factor receptor
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,05% proclin 300
- Molecular weight:90/140 kDa
- Sequence:CPTSCKCSASRIWCSDPSPGIVAFPRLEPNSVDPENITEIFIANQKRLEIINEDDVEAYVGLRNLTIVDSGLKFVAHKAFLKNSNLQHINFTRNKLTSLSRKHFRHLDLSELILVGNPFTCSCDIMWIKTLQEAKSSPDTQDLYCLNESSKNIPLANLQIPNCGLPSANLAAPNLTVEEGKSITLSCSVAGDPVPNMYWDVGNLVSKHMNETSHTQGSLRITNISSDDSGKQISCVAENLVGEDQDSVNLTVHFAPTITFLESPTSDHHWCIPFTVKGNPKPALQWFYNGAILNESKYICTKIHVTNHTEYHGCLQLDNPTHMNNGDYTLIAKNEYGKDEKQISAHFMGWPGIDDGANPNYPDVIYEDYGTAANDIGDTTNRSNEIPSTDVTDKTGREH
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 32 - 430 of human TrkB (NP_001018074.1).
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [ARC53486] antibody to TrkB for WB with samples derived from mouse and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:2000 to 1:10000
Type: Primary
Antigen: TrkB
Clonality: Monoclonal
Clone: ARC53486
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat