Anti-p53 (phospho Thr81) Rabbit Polyclonal Antibody
Catalog # ANTIA305390-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antigen NY-CO-13
- Antigen symbol:p53
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P04637
- Antigen synonyms:P53_HUMAN|Tp53|BCC7|Cellular tumor antigen p53|Mutant tumor protein 53|LFS1|Phosphoprotein p53|FLJ92943|p53 tumor suppressor
- Modification:phospho Thr81
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% proclin 300
- Molecular weight:53 kDa
- Sequence:APTPA
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic phosphorylated peptide around T81 of human TP53 (NP_000537.3).
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to p53 (phospho Thr81) for WB with samples derived from human and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:100 to 1:500
Type: Primary
Antigen: p53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat