Order Entry
Export
ContactUsLinkComponent
 
Anti-HSP90 alpha Mouse Monoclonal Antibody [clone: 2G5.G3]
  ANTIA305031-200
New Product
undefined
Anti-HSP90 alpha Mouse Monoclonal Antibody [clone: 2G5.G3]
  ANTIA305031-200
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    EL52
  • Antigen symbol:
    HSP90 alpha
  • Clonality:
    Monoclonal
  • Clone:
    2G5.G3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P07900
  • Antigen synonyms:
    Heat shock 86 kDa|heat shock 90kD protein, alpha-like 4|epididymis luminal secretory protein 52|heat shock 90kD protein 1, alpha|HS90A_HUMAN|Heat shock 90kDa protein 1 alpha|Heat shock protein HSP 90-alpha|Heat shock 90kD protein 1, alpha like 4|Heat shock protein 90kDa alpha (cytosolic) class A member 1
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,2, with 50% glycerol and 0,09% sodium azide
  • Molecular weight:
    ~90 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full length human HSP90 alpha protein.
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Protein G purification
  • Pack type:
    Plastic vial
  • Pk:
    200 µG

 

 

Mouse monoclonal [2G5.G3] antibody to HSP90 alpha for WB, IHC, ICC/IF, IP and ELISA with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF, IP, ELISA
  • Recommended dilutions: WB: 1:2000, ICC/IF: 1:100

Type: Primary
Antigen: HSP90 alpha
Clonality: Monoclonal
Clone: 2G5.G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat