Anti-AMIGO1 Mouse Monoclonal Antibody [clone: S86-36]
Catalog # ANTIA305013-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Adhesion molecule with Ig like domain 1
- Antigen symbol:AMIGO1
- Clonality:Monoclonal
- Clone:S86-36
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q80ZD8
- Antigen synonyms:AMIGO protein|AMIGO|AMIGO1|Alivin 2|ali2|Alivin-2|AMGO1_HUMAN|AMIGO-1|AMIGO 1
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
- Molecular weight:~60 - 80 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein amino acids 394 - 492 (cytoplasmic C - terminus) of mouse AMIGO - 1.
- Tested applications:IHC, ICC/IF
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [S86-36] antibody to AMIGO1 for WB, IHC and ICC/IF with samples derived from human, mouse and rat.
- Validated applications: WB, IHC, ICC/IF
- Recommended dilutions: WB: 1:1000
Type: Primary
Antigen: AMIGO1
Clonality: Monoclonal
Clone: S86-36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat