Order Entry
Export
ContactUsLinkComponent
Anti-SPATA2L Mouse Monoclonal Antibody [clone: Z2 P2D5-C8]
Anti-SPATA2L Mouse Monoclonal Antibody [clone: Z2 P2D5-C8]
Catalog # ANTIA304889-100
Supplier:  Antibodies.com
New Product
undefined
Anti-SPATA2L Mouse Monoclonal Antibody [clone: Z2 P2D5-C8]
Catalog # ANTIA304889-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C16orf76
  • Antigen symbol:
    SPATA2L
  • Clonality:
    Monoclonal
  • Clone:
    Z2 P2D5-C8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q8IUW3
  • Antigen synonyms:
    MGC26885|spermatogenesis associated 2 like|SPA2L_HUMAN|Spermatogenesis associated protein 2 like protein |SPATA2-like protein|SPATA2 like protein|chromosome 16 open reading frame 76|SPATA2L|Spermatogenesis-associated protein 2-like protein
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
  • Molecular weight:
    50 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide of Human SPATA2L.
  • Tested applications:
    IHC
  • Purification:
    Protein G purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [Z2 P2D5-C8] antibody to SPATA2L for WB, IHC and ELISA with samples derived from human.

  • Validated applications: WB, IHC, ELISA
  • Recommended dilutions: WB: 1:1000

Type: Primary
Antigen: SPATA2L
Clonality: Monoclonal
Clone: Z2 P2D5-C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human