Order Entry
Export
ContactUsLinkComponent
Anti-HSC70 Rabbit Polyclonal Antibody
Anti-HSC70 Rabbit Polyclonal Antibody
Catalog # ANTIA304835-100
Supplier:  Antibodies.com
New Product
undefined
Anti-HSC70 Rabbit Polyclonal Antibody
Catalog # ANTIA304835-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    2410008N15Rik
  • Antigen symbol:
    HSC70
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Fish,
    Salmon
  • Western blot:
    Yes
  • Form:
    lyophilised
  • Gene ID:
    UniprotID# B5DFX7
  • Antigen synonyms:
    Epididymis luminal protein 33|Heat shock 70kDa protein 8|Heat shock cognate protein 54|Epididymis secretory sperm binding protein Li 72p|Heat shock 70kD protein 8|Heat shock 70 kDa protein 8|Heat shock 70kD protein 10|Heat shock cognate 71 kDa protein|Constitutive heat shock protein 70
  • Storage buffer:
    Supplied as lyophilised antiserum
  • Molecular weight:
    ~73 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide conjugated to KLH. The peptide target is specific to the HSC70 of salmonid fish
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to HSC70 for WB with samples derived from fish and salmon.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:1000

Type: Primary
Antigen: HSC70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Fish;Salmon