Anti-SUR1 Mouse Monoclonal Antibody [clone: S289-16]
ANTIA304817-100
New Product
- Antibody type:Primary
- Antigen name:ABC36
- Antigen symbol:SUR1
- Clonality:Monoclonal
- Clone:S289-16
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Hamster,Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q09429
- Antigen synonyms:ATP-binding cassette sub-family C member 8|ATP binding cassette transporter sub family C member 8 (1)|Abcc8|ABCC8_HUMAN|MRP8|HRINS|ATP binding cassette sub family C (CFTR/MRP) member 8|HI|HHF1
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
- Molecular weight:~160 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein amino acids 1548 - 1582 (cytoplasmic C - terminus) of rat SUR1.
- Tested applications:IHC, ICC/IF
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [S289-16] antibody to SUR1 for WB, IHC and ICC/IF with samples derived from human, mouse, rat and hamster.
- Validated applications: WB, IHC, ICC/IF
- Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100
Type: Primary
Antigen: SUR1
Clonality: Monoclonal
Clone: S289-16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat;Hamster