Anti-GABA B Receptor 1 Mouse Monoclonal Antibody [clone: S93A-49]
Catalog # ANTIA304759-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:dJ271M21.1.1
- Antigen symbol:GABA B Receptor 1
- Clonality:Monoclonal
- Clone:S93A-49
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9Z0U4
- Antigen synonyms:GABABR1|GABA-B receptor 1|GABA B Receptor 1|GABAB subunit 1c|FLJ92613|GABA-B-R1|GABAB R1|dJ271M21.1.2|GABA-BR1
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
- Molecular weight:~115 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein amino acids 873 - 977 (cytoplasmic C - terminus) of rat GABA(B)R1.
- Tested applications:ICC/IF
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [S93A-49] antibody to GABA B Receptor 1 for WB and ICC/IF with samples derived from human, mouse and rat.
- Validated applications: WB, ICC/IF
- Recommended dilutions: WB: 1:1000
Type: Primary
Antigen: GABA B Receptor 1
Clonality: Monoclonal
Clone: S93A-49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat