Order Entry
Export
ContactUsLinkComponent
Anti-Chicken IgY H&L Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-Chicken IgY H&L Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA296105-1
Supplier:  Antibodies.com
New Product
Anti-Chicken IgY H&L Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA296105-1
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    Chicken IgY H&L
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Chicken
  • Form:
    Liquid
  • Antibody fragment:
    Whole Molecule
  • Storage buffer:
    Supplied in phosphate buffered saline with 0,2% BSA and 0,09% sodium azide
  • Storage temperature:
    Store in the dark at 4 °C. Avoid prolonged exposure to light. Do not freeze.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    IHC, ICC
  • Purification:
    Antigen affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    1 mg

Specifications

About this item

Goat anti-chicken IgY H&L (FITC) secondary antibody for IHC, ICC, Flow cytometry and IF with samples derived from chicken.

  • Validated applications: IHC, ICC, Flow cytometry, IF
  • Recommended dilutions: Flow cytometry: 1:50 to 1:200, ICC: 1:50 to 1:500, IF: 1:50 to 1:500, IHC: 1:50 to 1:500

Type: Secondary
Antigen: Chicken IgY H&L
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Chicken