Order Entry
Export
ContactUsLinkComponent
Anti-Rat IgM Goat Polyclonal Antibody (Biotin)
Anti-Rat IgM Goat Polyclonal Antibody (Biotin)
Catalog # ANTIA294702-500
Supplier:  Antibodies.com
New Product
Anti-Rat IgM Goat Polyclonal Antibody (Biotin)
Catalog # ANTIA294702-500
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    Rat IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antibody fragment:
    Whole Molecule
  • Storage buffer:
    Supplied in phosphate buffered saline with 0,2% BSA and 0,09% sodium azide
  • Storage temperature:
    Shipped at 4 °C. Store at 4 °C. Do not freeze.
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    IHC, ICC
  • Purification:
    Antigen affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    500 µG

Specifications

About this item

Goat anti-rat IgM (Biotin) secondary antibody for WB, IHC, ICC and ELISA with samples derived from rat.

  • Validated applications: WB, IHC, ICC, ELISA
  • Recommended dilutions: ELISA: 1:10000 to 1:200000, ICC: 1:100 to 1:500, IHC: 1:250 to 1:2500, WB: 1:10000 to 1:200000

Type: Secondary
Antigen: Rat IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat