Order Entry
Export
ContactUsLinkComponent
Anti-EGFR (phospho Tyr1068) Rabbit Recombinant Antibody [clone: RM443]
Anti-EGFR (phospho Tyr1068) Rabbit Recombinant Antibody [clone: RM443]
Catalog # ANTIA290827-100
Supplier:  Antibodies.com
New Product
Anti-EGFR (phospho Tyr1068) Rabbit Recombinant Antibody [clone: RM443]
Catalog # ANTIA290827-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    9030024J15Rik
  • Antigen symbol:
    EGFR
  • Clonality:
    Recombinant
  • Clone:
    RM443
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P00533
  • Antigen synonyms:
    EGFR_HUMAN|EGF R|Cell growth inhibiting protein 40 |EGFR|Cell proliferation inducing protein 61 |Avian erythroblastic leukemia viral (v erb b) oncogene homolog |AI552599|Epidermal growth factor receptor|Avian erythroblastic leukemia viral (verbb) oncogene homolog
  • Modification:
    phospho Tyr1068
  • Storage buffer:
    Supplied in phosphate buffered saline with 50% glycerol, 1% BSA, and 0,09% sodium azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic phospho - peptide corresponding to human EGFR (phospho Tyr1068).
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Recombinant rabbit monoclonal [RM443] antibody to EGFR (phospho Tyr1068) for WB with samples derived from human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:1000 to 1:4,000

Type: Primary
Antigen: EGFR
Clonality: Recombinant
Clone: RM443
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human