Order Entry
Export
ContactUsLinkComponent
Anti-IRF4 Goat Polyclonal Antibody
Anti-IRF4 Goat Polyclonal Antibody
  ANTIA286064-100
New Product
Anti-IRF4 Goat Polyclonal Antibody
  ANTIA286064-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Interferon regulatory factor 4
  • Antigen symbol:
    IRF4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    3662, 16364, 291100
  • Antigen synonyms:
    IRF4_HUMAN|Lymphocyte specific IRF|Lymphocyte specific interferon regulatory factor|IRF-4|IRF 4|LSIRF|Irf4|Multiple myeloma oncogene 1|Lymphocyte-specific interferon regulatory factor
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,3, with 0,02% sodium azide and 0,5% BSA
  • Sequence:
    C-DPQRQRKLITAHVEP
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to the internal region of human IRF4.
  • Tested applications:
    IHC
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Goat polyclonal antibody to IRF4 for ELISA, WB, IHC and Flow cytometry with samples derived from human and rat.

  • Validated applications: ELISA, WB, IHC, Flow cytometry

Type: Primary
Antigen: IRF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Rat