Anti-KIR2DL4 Mouse Monoclonal Antibody [clone: mAb#33]
Catalog # ANTIA285844-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD158 antigen like family member D
- Antigen symbol:KIR2DL4
- Clonality:Monoclonal
- Clone:mAb#33
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q99706
- Antigen synonyms:CD158d|KI2L4_HUMAN|CD158 antigen-like family member D|CD_antigen=CD158d|Killer cell inhibitory receptor 103AS|Killer cell immunoglobulin like receptor 2DL4|Killer cell immunoglobulin-like receptor 2DL4|Killer cell immunoglobulin like receptor, two domains, long cytoplasmic tail, 4|G9P
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, without sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:NK3.3 cells and KIR2DL4 - Ig fusion protein.
- Tested applications:ICC, Functional Studies
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [mAb#33] antibody to KIR2DL4 for WB, ICC, functional studies, Flow cytometry and IP with samples derived from human.
- Validated applications: WB, ICC, Functional Studies, Flow cytometry, IP
- Recommended dilutions: Flow cytometry: 2 to 6 µg/ml
Type: Primary
Antigen: KIR2DL4
Clonality: Monoclonal
Clone: mAb#33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human