Anti-L-PLUNC Rabbit Recombinant Antibody [clone: LPLUNC1/7059R]
ANTIA278683-100
New Product
- Antibody type:Primary
- Antigen symbol:L-PLUNC
- Clonality:Recombinant
- Clone:LPLUNC1/7059R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q8TDL5
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:25 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full length human L - PLUNC protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [LPLUNC1/7059R] antibody to L-PLUNC for ELISA, WB and IHC-P with samples derived from human.
- Validated applications: ELISA, WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: L-PLUNC
Clonality: Recombinant
Clone: LPLUNC1/7059R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human